Learn More
Hach Company ORP Sensor 3/4in. Convertible
Catalog No. 5000207149 Shop All Hach Company Products
Encompass
3/4 Inch ORP Sensor-Convertible Style-PPS Body Material-Platinum Electrode Type-No Temperature Compensation-15 ft CablePPS Convertible Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation
Catalog No. 50-002-07149 Supplier Hach Company Supplier No. RC1R5N
3/4 Inch ORP Sensor-Convertible Style-PPS Body Material-Platinum Electrode Type-No Temperature Compensation-15 ft CablePPS Convertible Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation