Learn More
Hach Company ORP Sensor 3/4in. Convertible
Catalog No. 5000207498 Shop All Hach Company Products
Encompass
in Combination ORP Sensor-Convertible Style-PPS Body-Platinum Electrode Type- HF-Resistant Glass with PEI-4 5 m CablePPS Convertible Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation HF-Resistant Glass
Catalog No. 50-002-07498 Supplier Hach Company Supplier No. RC1R5NHF
in Combination ORP Sensor-Convertible Style-PPS Body-Platinum Electrode Type- HF-Resistant Glass with PEI-4 5 m CablePPS Convertible Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation HF-Resistant Glass