Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP Standard, 200 mV vs. Ag/AgCl, Reagents

Catalog No. 01334589
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-589 10 L
01-334-588 1 L
01-334-586 4 L
01-334-585 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-334-589 Supplier Reagents Holdings Llc Supplier No. CO00020010F
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ORP Standard, 200 mV vs. Ag/AgCl, Reagent, use for Redox. Store in a Cool Dry Place. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Standard, 200 mV vs. Ag/AgCl
Packaging Cubitainer
CAS 7732-18-5, 7447-40-7, 14459-95-1
Specific Gravity 1.0052
Physical Form Liquid
Technique Redox
Color Yellow/Green
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 10 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.