Learn More
Hach Company ORP Standard, Light's Solution Ampules, 20 mL, pk/25
Supplier:  Hach Company 2612520
Encompass
For checking electrode response in oxidation-reduction potential (ORP) measurements.
Catalog No. 50-199-6343