Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORP Standard, 250 mV vs. Ag/AgCl, Reagents

Catalog No. 01334584
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-584 4 L
01-334-583 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-584 Supplier Reagents Holdings Llc Supplier No. CO0002504BP
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ORP Standard, 250 mV vs. Ag/AgCl, Reagent, use for Redox. Recommended Storage: 2°C to 8°C (36°F to 46°F)Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Standard, 250 mV vs. Ag/AgCl
Packaging Opaque HDPE
CAS 7732-18-5, 7447-40-7, 14459-95-1
Specific Gravity 1.0052
Physical Form Liquid
Technique Redox
Color Yellow/Green
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 4 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.