Learn More
Hach Company ZoBell's ORP/Redox Standard Solution, 500 mL
Supplier:  Hach Company 2316949
Encompass
Standard for calibrating and checking accuracy of ORP/Redox electrodes. The ZoBell's ORP/Redox Standard Solution exhibits temperature correlation between 0 - 30°C. The standard value is dependent on the reference electrode type.
Catalog No. 50-199-5784