Learn More
Grainger ORP Transmitter Blind Ryton PPS
Catalog No. 502768171
Encompass_Preferred
The SENSOREX EM802 ORP ORP Transmitter is an industrial sensor that measures oxidation reduction potential in water treatment applications. It has a measurement range of -2000 to +2000 mV with an accuracy of ±5 mV. The sensor uses a platinum measuring electrode and silver/silver chloride reference electrode. It has an IP65 rated enclosure and 4-20 mA analog output. The EM802 is designed for continuous monitoring and control of ORP in municipal and industrial water treatment systems. It requires minimal maintenance.
Catalog No. 50-276-8171 Supplier Grainger Supplier No. 16Z036
The SENSOREX EM802 ORP ORP Transmitter is an industrial sensor that measures oxidation reduction potential in water treatment applications. It has a measurement range of -2000 to +2000 mV with an accuracy of ±5 mV. The sensor uses a platinum measuring electrode and silver/silver chloride reference electrode. It has an IP65 rated enclosure and 4-20 mA analog output. The EM802 is designed for continuous monitoring and control of ORP in municipal and industrial water treatment systems. It requires minimal maintenance.