Learn More
Sino Biological ORP150 / HYOU1 / HSP12A Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11342RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HSP12A (rh HSP12A; Catalog#11342-H08H; NP_001124463.1; Met 695-Leu 994). HSP12A specific IgG was purified by human HSP12A affinity chromatography.
Catalog No. 50-224-8619