Learn More
Sino Biological ORP150 / ORP150 / HYOU1 / HSP12A Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11342R201100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ORP150 / HYOU1 / HSP12A (rh ORP150 / HYOU1 / HSP12A; Catalog#11342-H08H; NP_001124463.1; Met 695-Leu 994).
Catalog No. 50-224-7793