Learn More
Cole-Parmer Oakton Replacement Sensor for ORPTestr 5
Catalog No. 501961771 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample. For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.
Catalog No. 50-196-1771 Supplier Cole-Parmer Supplier No. 3563458
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample. For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.