Learn More
Cole-Parmer Oakton ORP electrode, epoxy body, single junction, 12 mm dia x 110 mm L, 0.9-m cable
Catalog No. 501961374 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1374 Supplier Cole-Parmer Supplier No. 3580513
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.