Learn More
Cole-Parmer Oakton ORP Electrode, Single-Junction, Submersible; 12 x 130 mm
Catalog No. 501961375 Shop All Cole-Parmer Products
Encompass_Preferred
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212>F (-5 to 100>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230>F (-5 to 110>C).
Catalog No. 50-196-1375 Supplier Cole-Parmer Supplier No. 3580514
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212>F (-5 to 100>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230>F (-5 to 110>C).