Learn More
Cole-Parmer Oakton ORP Electrode, Epoxy, Single-Junction; 10 ft cable
Catalog No. 501961809 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1809 Supplier Cole-Parmer Supplier No. 3580525
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.