Learn More
CPC Scientific H-Ala-Ser-Gly-Pro-Leu-Gly-Thr-Leu-Ala-Glu-Glu-Leu-Ser-Ser-Tyr-Ser-Arg-Arg-Lys-Gly-Gly-Phe-Ser-Phe-Arg-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OPIO015B
Encompass
P550 (mouse) is the mouse homolog of P518 (human). It has been shown to exhibit similar agonistic activity towards the human orphan G-protein-coupled receptor SP9155 as P518 (human) (EC50 = 6 nM).ONE-LETTER SEQUENCE: ASGPLGTLAEELSSYSRRKGGFSFRF-NH2MOLECULAR FORMULA: C126H195N37O37MOLECULAR WEIGHT:2820.16STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [600171-70-8], RESEARCH AREA: PainREFERENCES: N.Chartrel et al., Proc. Natl. Acad. Sci. USA, 100, 15247 (2003); Y.Jiang et al., J. Biol. Chem., 278, 27652 (2003)
Catalog No. 50-210-9992