Learn More
Enzo Life Sciences SCREEN-WELL Orphan ligand library (500 µl/well)
SDP
Supplier:  Enzo Life Sciences BML28250500
Encompass_Preferred
The SCREEN-WELL Orphan ligand library contains 84 compounds with defined, putative, potential or speculative biological activity but whose protein binding partners have not been identified. Ligands are supplied dissolved in biocompatible solvents at 10mM and aliquoted into a deep-well plate at 500µl per well. The library is a rich source of ligands for receptor de-orphaning. Includes the following classes of ligands: Trace Amines, Neurotransmitter Metabolites, Endogenous β-carbolines, Urinary Metabolites, Nicotine Congeners, D-Amino Acids. Receptors for endogenous trace amines have been deorphanized. Endogenous β-carbolines have been shown to display varied biological effects such as hallucinations, hypotension and others. Nicotine congeners such as nornicotine are biologically active and may act at novel nicotinic receptor subtypes. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0208