Learn More
Apexbio Technology LLC Orphenadrine Citrate 4682-36-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090595
Encompass_Preferred
Orphenadrine Citrate (CAS 4682-36-4) is a small molecule utilized in biomedical research for its pharmacological modulation of neurological pathways It acts by inhibiting the binding of [ 3H]MK-801 to the phencyclidine (PCP) site of the N-methyl-D-aspartate (NMDA) receptor in human frontal cortex tissue implicating antagonism at NMDA-type glutamate receptors Orphenadrine has historically served as an antiparkinsonian antispastic and analgesic agent Its ability to modulate NMDA receptor activity makes it a valuable tool for studies investigating glutamatergic neurotransmission and related neuropharmacological mechanisms
Catalog No. 50-000-90595 Supplier Apexbio Technology LLC Supplier No. B16065.1
Add to Cart
Add to Cart
Orphenadrine Citrate (CAS 4682-36-4) is a small molecule utilized in biomedical research for its pharmacological modulation of neurological pathways It acts by inhibiting the binding of [ 3H]MK-801 to the phencyclidine (PCP) site of the N-methyl-D-aspartate (NMDA) receptor in human frontal cortex tissue implicating antagonism at NMDA-type glutamate receptors Orphenadrine has historically served as an antiparkinsonian antispastic and analgesic agent Its ability to modulate NMDA receptor activity makes it a valuable tool for studies investigating glutamatergic neurotransmission and related neuropharmacological mechanisms