Learn More
Apexbio Technology LLC Orphenadrine Citrate 4682-36-4 50mg
SDP
Catalog No. 502240698
Encompass_Preferred
Orphenadrine Citrate (CAS 4682-36-4) is a small molecule utilized in biomedical research for its pharmacological modulation of neurological pathways It acts by inhibiting the binding of [ 3H]MK-801 to the phencyclidine (PCP) site of the N-methyl-D-aspartate (NMDA) receptor in human frontal cortex tissue implicating antagonism at NMDA-type glutamate receptors Orphenadrine has historically served as an antiparkinsonian antispastic and analgesic agent Its ability to modulate NMDA receptor activity makes it a valuable tool for studies investigating glutamatergic neurotransmission and related neuropharmacological mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0698 Supplier Apexbio Technology LLC Supplier No. B160650
Orphenadrine Citrate (CAS 4682-36-4) is a small molecule utilized in biomedical research for its pharmacological modulation of neurological pathways It acts by inhibiting the binding of [ 3H]MK-801 to the phencyclidine (PCP) site of the N-methyl-D-aspartate (NMDA) receptor in human frontal cortex tissue implicating antagonism at NMDA-type glutamate receptors Orphenadrine has historically served as an antiparkinsonian antispastic and analgesic agent Its ability to modulate NMDA receptor activity makes it a valuable tool for studies investigating glutamatergic neurotransmission and related neuropharmacological mechanisms