Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orphenadrine citrate salt, MilliporeSigma™ Supelco™

Catalog No. 111014235 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4235 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4235 Supplier MilliporeSigma Supelco Supplier No. O46305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 4682-36-4
Physical Form Neat
Linear Formula C18H23 NO · C6H8O7
MDL Number MFCD00079197
Synonym beta-Dimethylaminoethyl 2-methylbenzhydryl ether citrate salt; o-Methyldiphenhydramine citrate salt
Type Analytical Standard
Quantity 5 g
Formula Weight 461.50
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.