Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Orphenadrine Citrate, USP
SDP

Catalog No. 14185665 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-665 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-665 Supplier Medisca Supplier No. 045209
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orphenadrine Citrate USP is a white, practically odorless, crystalline powder.

Orphenadrine Citrate USP is a white, practically odorless, crystalline powder.

Specifications

Chemical Name or Material Orphenadrine Citrate, USP
Packaging Jar 3800ml, White, HDPE, Round, 110-400, TRIVENI-DB, CT
CAS 4682-36-4
Type Drug Standard
Molecular Weight (g/mol) 461.5
Quantity 1 kg
Percent Purity 98 to 101.5 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.