Learn More
TARGETMOL CHEMICALS INC Orphenadrine hydrochloride
SDP
Catalog No. 502880468
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Orphenadrine hydrochloride (Mephenamin) is a muscarinic antagonist used to treat drug-induced parkinsonism and to relieve pain from muscle spasm. Purity 99.54%
Catalog No. 50-288-0468 Supplier TARGETMOL CHEMICALS INC Supplier No. T1308500MG
Also available in 1 g, 1 mL, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Orphenadrine hydrochloride (Mephenamin) is a muscarinic antagonist used to treat drug-induced parkinsonism and to relieve pain from muscle spasm. Purity 99.54%