Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orphenadrine Hydrochloride 99%, Thermo Scientific™

Catalog No. AC157500100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157500100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC157500100 Supplier Thermo Scientific Chemicals Supplier No. 157500100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341-69-5
Molecular Formula C18H24ClNO
Molecular Weight (g/mol) 305.846
InChI Key UQZKYYIKWZOKKD-UHFFFAOYSA-N
Synonym orphenadrine hydrochloride, orphenadrine hcl, mebedrol, mephenamin, mefenamin hydrochloride, mephenamin forte, mephenamin hydrochloride, orfenadrin hydrochloride, mephenamine hydrochloride
PubChem CID 9568
ChEBI CHEBI:60902
IUPAC Name N,N-dimethyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
SMILES CC1=CC=CC=C1C(C2=CC=CC=C2)OCCN(C)C.Cl

Specifications

Melting Point 159°C to 162°C
Color White
Infrared Spectrum Authentic
Linear Formula CH3C6H4CH(C6H5)OCH2CH2N(CH3)2.HCl
Quantity 10 g
Merck Index 14, 6879
Formula Weight 305.84
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Orphenadrine hydrochloride, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.