Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orphenadrine Hydrochloride, TCI America™
SDP

Catalog No. O040625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O040625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O040625G Supplier TCI America Supplier No. O040625G

Specifications

CAS 341-69-5
Melting Point 161°C
Color White
Molecular Formula C18H24ClNO
MDL Number MFCD00012480
UN Number 2811
Quantity 25 g
Synonym orphenadrine hydrochloride, orphenadrine hcl, mebedrol, mephenamin, mefenamin hydrochloride, mephenamin forte, mephenamin hydrochloride, orfenadrin hydrochloride, mephenamine hydrochloride
InChI Key UQZKYYIKWZOKKD-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1C(C2=CC=CC=C2)OCCN(C)C.Cl
IUPAC Name N,N-dimethyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
Molecular Weight (g/mol) 305.846
PubChem CID 9568
ChEBI CHEBI:60902
Formula Weight 305.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Orphenadrine Hydrochloride
Show More Show Less
RTECSNumber KR6300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.