Learn More
Cayman Chemical Orsellinic Acid, Purity: ≥98%, CAS: 480-64-8, 10 mg
SDP
Catalog No. 502995612
Encompass_Preferred
A fungal metabolite and benzoic acid derivative with antioxidant and neuroprotective activities; scavenges DPPH radicals (IC50 = 5 mM); prevents PAF-induced PARP cleavage in PC12-AC cells and PAF-induced cytotoxicity in Pafr-/-mouse cerebellar granule cells at 1 μg/ml
Catalog No. 50-299-5612 Supplier Cayman Chemical Supplier No. 2270810 MG
A fungal metabolite and benzoic acid derivative with antioxidant and neuroprotective activities; scavenges DPPH radicals (IC50 = 5 mM); prevents PAF-induced PARP cleavage in PC12-AC cells and PAF-induced cytotoxicity in Pafr-/-mouse cerebellar granule cells at 1 μg/ml