Learn More
Selleck Chemical LLC Orteronel S1195-5mg
SDP
Catalog No. 503108244
Encompass_Preferred
Orteronel(TAK-700) is a potent and highly selective human 17 20-lyase inhibitor with IC50 of 38 nM exhibits 1000-fold selectivity over other CYPs (e g 11-hydroxylase and CYP3A4) TAK-700 (Orteronel) is an androgen biosynthesis inhibitor Phase 3
Catalog No. 50-310-8244 Supplier Selleck Chemical LLC Supplier No. S11955MG
Orteronel(TAK-700) is a potent and highly selective human 17 20-lyase inhibitor with IC50 of 38 nM exhibits 1000-fold selectivity over other CYPs (e g 11-hydroxylase and CYP3A4) TAK-700 (Orteronel) is an androgen biosynthesis inhibitor Phase 3