Learn More
Medchemexpress LLC HY-10505 5mg Medchemexpress, Orteronel CAS:566939-85-3 Purity:>98%
SDP
Catalog No. 501968245
Encompass_Preferred
Medchemexpress, HY-10505 5mg Orteronel CAS:566939-85-3 Orteronel is a highly selective inhibitor of human 17,20-lyase with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8245 Supplier Medchemexpress LLC Supplier No. HY105055MG
Medchemexpress, HY-10505 5mg Orteronel CAS:566939-85-3 Orteronel is a highly selective inhibitor of human 17,20-lyase with IC50 of 38 nM, and exhibits >1000-fold selectivity over other CYPs such as 11-hydroxylase and CYP3A4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.