Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho ORTHO 7% BSA
SDP

Catalog No. 21377803 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Concentration:
7%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
21-377-803 7%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-377-803 Supplier QuidelOrtho Supplier No. 6844285
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An aqueous solution of bovine serum, inorganic salts, and preservatives.

Aqueous solution consisting of:

  • bovine serum
  • inorganic salts
  • preservatives

Specifications

CE Marker Yes
Concentration 7%
Type BSA Solution
Formulation 7% BSA is an aqueous solution of bovine serum, inorganic salts, and preservatives.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.