Learn More
QuidelOrtho ABO Blood Group Reagents
SDP
Catalog No. p-109904
Change view
Click to view available options
Quantity:
3 x 10 mL
15 x 10 mL
Type:
Anti-A BioClone™
Anti-A, B BioClone™
Anti-B BioClone™
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-287248 15 x 10 mL Anti-B BioClone™
23-287247 15 x 10 mL Anti-A BioClone™
23-287249 15 x 10 mL Anti-A, B BioClone™
23-287-450 3 x 10 mL Anti-A BioClone™
23-287-451 3 x 10 mL Anti-B BioClone™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.
Catalog No. 23-287248 Supplier QuidelOrtho Supplier No. 711320
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.
Anti-A, Anti-B and Anti-A,B reagents are used to perform ABO confirmations
  • Anti-A and Anti-B BioClone™ reagents are used to determine ABO cell type
  • Provide the strong reactivity and product consistency associated with monoclonal technology
  • Reagents are Murine Monoclonal blends
  • Anti-A,B BioClone reagent is a blend of Anti-A and Anti-B monoclonal antibodies used for ABO group confirmation; can be used by transfusion services to confirm blood group O donor units
  • Anti-A and Anti-A,B can detect many A subgroups (such as A3, Ax, or AsubB)
Specifications
Type Anti-B BioClone™
For Use With (Application) To determine ABO cell type
Quantity 15 x 10 mL