Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho BioClone™ Reagents
SDP

Catalog No. OK713080 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 5 mL
3 x 10 mL
15 x 10 mL
Type:
Anti-C (Anti-RH2) BioClone™ Reagent
Anti-D BioClone™ Reagent
Anti-E (Anti-RH3) BioClone™ Reagent
Anti-c BioClone™ Reagent
Anti-e BioClone™ Reagent
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
OK713080 1 x 5 mL Anti-C (Anti-RH2) BioClone™ Reagent
23-287-320 1 x 5 mL Anti-E (Anti-RH3) BioClone™ Reagent
23-287-321 1 x 5 mL Anti-e BioClone™ Reagent
23-287402 15 x 10 mL Anti-D BioClone™ Reagent
23-287-452 3 x 10 mL Anti-D BioClone™ Reagent
23-287-453 1 x 5 mL Anti-c BioClone™ Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.
Catalog No. OK713080 Supplier QuidelOrtho Supplier No. 713080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.

Anti Sera products provide qualitative tests for recognition of antigens on human red blood cells

  • Using high-protein reagents prepared from human serum, Anti-D blood grouping reagent recognizes D (Rh0) antigens
  • Anti-C (Anti-RH2), Anti-E (Anti-RH3), Anti-c (Anti-RH4) and Anti-e (Anti-RH5) blood grouping reagents are low-protein reagents prepared from human/mouse cell lines; used to recognize corresponding antigens on human red blood cells
The Rh-hr control reagent is designed to recognize spontaneous agglutination of red blood cells
  • The control is prepared from pools of human serum tested and found to be negative for blood group antibodies

Specifications

Type Anti-C (Anti-RH2) BioClone™ Reagent
For Use With (Application) To recognize spontaneous Agglutination of red Blood cells
Quantity 1 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.