Learn More
QuidelOrtho Anti-D Sera and Rh-hr Typing Reagents: QuidelOrtho Rh-hr Control
Shop All Ortho-Clinical Diagnostics Products
Description
The Rh-hr control reagent is designed to recognize spontaneous agglutination of red blood cells
- The control is prepared from pools of human serum tested and found to be negative for blood group antibodies
Specifications
Specifications
| Type | Rh-hr Control Reagent |
| For Use With (Application) | To recognize spontaneous Agglutination of red Blood cells |
| Quantity | 15 x 10 mL |