Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Antihuman Globulin Reagents
SDP

Catalog No. 23287244 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 3 mL
1 x 10 mL
15 x 10 mL
100 x 10 mL
Type:
Anti-C3d (Murine Monoclonal) Reagent
Anti-IgG (Rabbit) Reagent
Anti-IgG,-C3d Reagent
Color:
Clear
Green
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Color
23-287244 1 x 10 mL Anti-IgG (Rabbit) Reagent Clear
OK716080 15 x 10 mL Anti-IgG,-C3d Reagent Clear
OK716530 1 x 3 mL Anti-C3d (Murine Monoclonal) Reagent Clear
23-252480 1 x 10 mL Anti-IgG (Rabbit) Reagent Green
23-287-454 100 x 10 mL Anti-IgG (Rabbit) Reagent Clear
23-287-455 100 x 10 mL Anti-IgG (Rabbit) Reagent Green
23-287-483 100 x 10 mL Anti-IgG,-C3d Reagent Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.
Catalog No. 23-287244 Supplier QuidelOrtho Supplier No. 716360
Add to Cart
Add to Cart
Contains sodium azide. Sodium azide may react with lead and copper plumbing to form highly explosive metal azide. On disposal, flush with a large volume of water to prevent azide buildup.

QuidelOrtho antihuman globulin reagents are used in direct and indirect antiglobulin tests

  • BioClone™ Anti-IgG, -C3d is used for indirect and direct antiglobulin testing
  • Manufactured using rabbit polyclonal Anti-IgG and monoclonal Anti-C3b and Anti-C3d
  • Available in clear or green
  • QuidelOrtho Anti-IgG is a monospecific reagent used for indirect antiglobulin testing for the detection and identification of antibodies
  • Also used in further characterization of proteins on red blood cells
  • Available in clear or green
  • Anti-C3d BioClone characterizes proteins present on red blood cells

Specifications

Color Clear
Type Anti-IgG (Rabbit) Reagent
For Use With (Application) For indirect antiglobulin testing for the detection and identification of Antibodies
Quantity 1 x 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.