Learn More
Description
- BioClone™ Anti-IgG, -C3d is used for indirect and direct antiglobulin testing
- Manufactured using rabbit polyclonal Anti-IgG and monoclonal Anti-C3b and Anti-C3d
- Available in clear or green
- QuidelOrtho Anti-IgG is a monospecific reagent used for indirect antiglobulin testing for the detection and identification of antibodies
- Also used in further characterization of proteins on red blood cells
- Available in clear or green
- Anti-C3d BioClone characterizes proteins present on red blood cells
Specifications
Specifications
| Color | Clear |
| Type | Anti-IgG,-C3d Reagent |
| For Use With (Application) | For indirect antiglobulin testing for the detection and identification of Antibodies |
| Quantity | 15 x 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
