Description
- Reagents are used to enhance antigen/antibody interactions
- Results in an increase in speed and sensitivity of detection
Specifications
Specifications
| Type | Bovine Serum Albumin Solution |
| For Use With (Application) | For Blood typing reactions, enhance Antigen/antibody interactions, increase in speed and sensitivity of detection |
| Quantity | 100 x 10 mL |