Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho BioClone™ Anti-IgG, -C3d Polyspecific Anti-Human Green Globulin Reagent
SDP

Catalog No. 23287245 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
15 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-287245 15 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-287245 Supplier QuidelOrtho Supplier No. 716980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal reagent for indirect and direct antiglobulin testing

  • Manufactured using rabbit polyclonal Anti-IgG and monoclonal Anti-C3b and Anti-C3d
  • Contains no Anti-C4 so additional problem solving caused by the presence of Anti-C4 is eliminated
  • Indirect test is used for antibody detection and identification, crossmatching, and antigen typing
  • Direct test detects presence of in vivo coating of red blood cells with IgG and/or the complement components C3b and/or C3d
  • Green color aids in visual inspection for addition of antihuman globulin in manual or automated procedures

Specifications

Type Anti-Human Globulin
For Use With (Application) For Antibody detection and identification, crossmatching and Antigen typing and indirect and direct antiglobulin testing
Quantity 15 x 10 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.