Learn More
QuidelOrtho Antihuman Globulin Anti-C3b, -C3d BioClone™ Reagent
SDP
Catalog No. 23287315 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 3 mL
Catalog No. Quantity
23-287-315 1 x 3 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-287-315 Supplier QuidelOrtho Supplier No. 716540
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determine presence or absence of complement components on human red blood cells that are agglutinated by polyspecific antihuman globulin
  • Quantity: 3mL/Pk.
Specifications
Type Anti-C3b, -C3d (Murine monoclonal) BioClone™
Quantity 1 x 3 mL