Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Anti-Fyb Reagents
SDP

Catalog No. 23287308 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-287-308 1 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-287-308 Supplier QuidelOrtho Supplier No. 725230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Prepared from human sera containing Fyb antibodies
  • Used in selecting donor red cells for patients who became Fyb antigen immune as result of transfusion or pregnancy
  • Quantity: 1 x 5mL

Specifications

Type QuidelOrtho Anti-Fyb Reagent
Quantity 1 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.