Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Anti-k (Anti-Cellano) Reagents
SDP

Catalog No. 23287311 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-287-311 1 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-287-311 Supplier QuidelOrtho Supplier No. 721030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For indirect antiglobulin test to recognize k-(Cellano) antigen on human red blood cells

  • Prepared from human sera containing K antibodies
  • Quantity: 1 x 3mL

Specifications

Type QuidelOrtho Anti-k (Anti-Cellano) Reagent
Quantity 1 x 3 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.