Learn More
QuidelOrtho Confidence System Blood Bank Controls
SDP
Catalog No. 23287303 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
23-287-303 Each
1 options
This item is not returnable. View return policy
Catalog No. 23-287-303 Supplier QuidelOrtho Supplier No. 6902096
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accurate and reliable quality control system for blood bank reagent of tube-based test systems
  • Antibody and reagent red cells provides means of confirming reactivity of reagents used to test for ABO, Rh and antibody detection test reagents
Specifications
Type QuidelOrtho Confidence System Blood Bank Control
For Use With (Application) To validate Blood bank procedures
Quantity Each