Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho RESOLVE™ Reagent Red Blood Cells: Panel B
SDP

Catalog No. 23287465 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Quantity:
11 x 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-287-465 11 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-287-465 Supplier QuidelOrtho Supplier No. 6902318
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For detection and identification of blood group antibodies

  • Prepared in 3% (for tube tests) and 0.8% (for ID-MTS™ brand Gel Test™) suspensions

RESOLVE Panel B:

  • Designed to complement RESOLVE Panel A and to serve as a selected cell panel
  • Allows identification of more difficult antibody combinations by providing specially selected cells
  • Presence of RzR1 and Cellano negative cell assured of every panel

Specifications

Type 8% Resolve™ Panel B
Quantity 11 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.