Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho RESOLVE™ Reagent Red Blood Cells (Panels): Panel A
SDP

Catalog No. 23287464 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Type:
3% Resolve™ Panel A
8% Resolve™ Panel A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-287-464 8% Resolve™ Panel A
23-287-302 3% Resolve™ Panel A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-287-464 Supplier QuidelOrtho Supplier No. 6902317
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For detection and identification of blood group antibodies

  • Prepared in 3% (for tube tests) and 0.8% (for ID-MTS™ brand Gel Test™) suspensions

RESOLVE Panel A:

  • 11-cell panel that provides selection of cells to assure identification of antibodies to common blood group factors, including those known to exhibit dosag
  • Cell 11 will represent unusual Rh phenotype, possess antigen of low incidence or lack antigen of high incidence

Specifications

Type 8% Resolve™ Panel A
Quantity 11 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.