Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ 4600 Chemistry System

Catalog No. 23057102
Change view
Click to view available options
No. of Channels:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Channels
23-057-102 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 23-057-102 Supplier QuidelOrtho Supplier No. 6802445
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Specifications

Depth (English) 33.4 in.
Product Type Chemistry System
Amperage 20 A
Operating Temperature Range (English) 59°F to 86°F
Depth (Metric) 84.8 cm
Operating Temperature Range (Metric) 15°C to 30°C
Volume (Metric) Sample 2 to 17 μL
Frequency 47/63 Hz
No. of Channels 10
Electrical Requirements 200 to 240V 47 to 63Hz
Holds 5, 7, 10mL collection tubes, Vitros™ microsample cups and 0.5 and 2mL cups
Data Storage CD-ROM/USB
Includes Color coded graphical user interface, ergonomic flat, low-glare, LCD, 17 in. touchscreen monitor, numeric keypad on monitor, detached keyboard for maximum flexibility, onboard documentation and animated help (V-Docs), onboard maintenance documentation
Test Time Potentiometric ≈2.5 min., colorimetric ≈6 min., immuno rate ≈9 min., MicroTip™ ≈8 to 16 min., for single result
Calibration Time 6 Months
Height (English) 52.5 in.
Height (Metric) 1.33 m
Humidity 15 to 75%
Sample Size 150 routine sample in universal sample tray, 10 samples in dedicated STAT lane
Sample Type Serum, Plasma, Urine, CSF, Whole Blood
Voltage 200/240 V
Weight (English) 1400 lb.
Weight (Metric) 640 kg
Width (English) 92.2 in.
Width (Metric) 234.1 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.