Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuidelOrtho VITROS™ B12/Folate Denaturing Reagent
Specifications
Specifications
| Detectable Analytes | Vitamin B12 |
| Sample Type | EDTA Plasma, Heparin Plasma, Serum |
| Type | B12/Folate Denaturing Reagent |
| For Use With (Application) | Folate deficiency, in-vitro diagnostic use only |
| Includes | 11.7mL Sample treatment (denaturation) reagent, 15.6mL sample treatment (neutralization) reagent, ancillary lot card |
| Quantity | 200 Tests |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?