Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ Calibrators: Anti-Hepatitis A virus Total

Catalog No. OCD6801462
Change view
Click to view available options
Quantity:
1 Vial/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD6801462 1 Vial/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD6801462 Supplier QuidelOrtho Supplier No. 6801462
Add to Cart
Add to Cart

Specifications

Format Vial
For Use With (Equipment) VITROS™ ECi immunodiagnostic system
Type Total anti-HAV Calibrator
Level 1
Quantity 1 Vial/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.