Learn More
QuidelOrtho VITROS™ Calibrators: Anti-Hepatitis B core Antigen IGM
Specifications
Specifications
| Format | Vial |
| For Use With (Equipment) | VITROS™ ECi immunodiagnostic system |
| Type | Anti-HBc IgM Calibrator |
| Intended Use | Qualitative detection of IgM Antibody to Hepatitis B core Antigen (Anti-HBc IgM) |
| Quantity | 1 x 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.