Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ Calibrators: Carcinoembryonic Antigen

Catalog No. OCD1062306
Change view
Click to view available options
Quantity:
1 Set/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD1062306 1 Set/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD1062306 Supplier QuidelOrtho Supplier No. 1062306
Add to Cart
Add to Cart

Specifications

Content And Storage 2°C to 8°C
For Use With (Equipment) VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system
Sample Type Human Serum and Plasma (EDTA or Heparin)
Type Carcinoembryonic Antigen (CEA) Calibrator
Includes 2mL VITROS™ CEA calibrators 1 and 2 (Human CEA in Bovine Serum with antimicrobial agent), lot calibration card, protocol card, 16 calibrator bar code labels (8 for each calibrator)
Intended Use Quantitative measurement of CEA
Level 3
Quantity 1 Set/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.