Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuidelOrtho VITROS™ Calibrators: Ferritin
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
| Sample Type | Human Serum and Plasma (Heparin) |
| Type | Ferritin Calibrator |
| Includes | VITROS™ Ferritin calibrators 1, 2 and 3 (freeze-dried, Human Spleen Ferritin in Human Plasma with antimicrobial agent, reconstitution volume 1mL), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) |
| Intended Use | Quantitative measurement of Ferritin |
| Level | 3 |
| Lyophilized | Yes |
| Quantity | 3 Sets/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?