Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ Calibrators: Folate

Catalog No. OCD1311075
Change view
Click to view available options
Quantity:
1 Set/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OCD1311075 1 Set/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OCD1311075 Supplier QuidelOrtho Supplier No. 1311075
Add to Cart
Add to Cart

Specifications

Content And Storage 2°C to 8°C
For Use With (Equipment) VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system
Sample Type Human Serum, Plasma (Heparin), Whole Blood
Type Folate Calibrator
Includes 2.1mL VITROS™ Folate calibrators 1, 2 and 3 (Folic Acid in Buffer with Human Serum Albumin and antimicrobial agent), lot calibration card (for use with reagent pack 1, colored green), ancillary lot card (for use with reagent pack 2, colo
Intended Use Quantitative measurement of Folate
Level 3
Quantity 1 Set/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.