Learn More
QuidelOrtho VITROS™ Calibrators: Follicle-stimulating Hormone
Description
The VITROS FSH detection kit is designed for the quantitative determination of follicle stimulating hormone (FSH) in human serum or plasma using a chemiluminescence immunoassay double sandwich method.
- Test Principle: Uses streptavidin magnetic particles (R1), acridinium ester-labeled FSH antibody (R2), and biotin-labeled FSH antibody (R3) to form an antigen-antibody complex.
- Analytical Range: 0.2 mIU/mL to 200 mIU/mL.
- Minimum Detection Limit: 0.1 mIU/mL.
- Performance Characteristics: Accuracy is within ±15% of the nominal value, repeatability is 8.0%, and between-batch difference is 15.0%.
- Sample Type: Serum or plasma.
- Compatibility: Suitable for use with CM Series Chemiluminescence Immunoassay Analyzers and CSM Series Integrated Systems.
- Storage: Store at 2°C to 8°C, airtight, upright, and protected from sunlight.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | FSH |
| For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
| Sample Type | Human Serum and Plasma (EDTA or Heparin) |
| Type | Follicle Stimulating Hormone (FSH) Calibrator |
| Includes | 2mL VITROS™ FSH calibrators 1 and 2 (Human Pituitary FSH in Bovine Serum with antimicrobial agent), lot calibration card, protocol card, 16 calibrator bar code labels (8 for each calibrator) |
| Intended Use | Quantitative measurement of Follicle Stimulating Hormone (FSH) |
| Level | 2 |
| Quantity | 1 Set/Pk. |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.