Learn More
QuidelOrtho VITROS™ Calibrators: Hepatitis B Surface Antigen
Description
The VITROS™ HBsAg Calibrator is intended for use in the calibration of the VITROS Immunodiagnostic System for the quantitative or qualitative determination of Hepatitis B surface antigen (HBsAg).
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | HBsAg |
| Format | Vial |
| For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
| Sample Type | Human Serum and Plasma |
| Type | Hepatitis B Surface Antigen (HBSAG) Calibrator |
| Includes | 2mL VITROS™ HBsAg calibrator (Human HBsAg ad subtype, inactivated in Buffer with Bovine Serum Albumin and antimicrobial agent, lot calibration card, protocol card, 8 calibrator bar code labels |
| Intended Use | Qualitative in-vitro detection of Hepatitis B Surface Antigen (HBsAg) |
| Quantity | 1 Vial/Pk. |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.