Learn More
QuidelOrtho VITROS™ Calibrators: N-terminal Pro-brain Natriuretic Peptide
Shop All Ortho-Clinical Diagnostics Products

Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
| Sample Type | Human Serum and Plasma (EDTA or Heparin) |
| Type | N-terminal Pro-Brain Natriuretic Peptide (NT-PROBNP) Calibrator |
| Includes | 2mL VITROS™ NT-proBNP calibrators 1, 2 and 3 (synthetic NT-pro-BNP in Buffer with Bovine Serum Albumin and antimicrobial agent), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) |
| Intended Use | Quantitative measurement of N-terminal pro Brain Natriuretic Peptide (NT-proBNP) |
| Level | 3 |
| Quantity | 1 Set/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.