Learn More
QuidelOrtho VITROS™ Calibrators: Prostate-specific antigen
Description
VITROS PSA Calibrator is intended for use in the calibration of the VITROS Immunodiagnostic Systems for the quantitative measurement of Prostate Specific Antigen (PSA) in human serum and plasma.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Prostate Specific Antigen (PSA) |
| For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
| Sample Type | Human Serum and Plasma (EDTA or Heparin) |
| Type | Prostate Specific Antigen (PSA) Calibrator |
| Includes | 1.75mL VITROS™ PSA calibrators 1, 2, and 3 (Human PSA in Buffer with antimicrobial agent and Bovine Serum Albumin), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) |
| Intended Use | Quantitative measurement of PSA |
| Level | 3 |
| Quantity | 1 Set/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.